missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Laurate, TRC

Product Code. 30408317
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30408317 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30408317 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCG61570550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Glycidyl Laurate
CAS 1984-77-6
Molekylformel C15 H28 O3
Kvantitet 50 mg
Synonym Dodecanoic Acid 2-Oxiranylmethyl Ester, Lauric Acid 2,3-Epoxypropyl Ester, 2,3-Epoxy-1-propanol Laurate, Glycidyl Dodecanoate, Lauric Acid Glycidyl Ester, NSC 406541
InChI formel InChI=1S/C15H28O3/c1-2-3-4-5-6-7-8-9-10-11-15(16)18-13-14-12-17-14/h14H,2-13H2,1H3
LEDER CCCCCCCCCCCC(=O)OCC1CO1
IUPAC-namn oxiran-2-ylmethyl dodecanoate
Molekylvikt (g/mol) 256.38
Formel vikt 256.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.