Läs mer
Glycidyl Linoleate, TRC
Produktkod. 17929834
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17929834 10 mg
17909174 25 mg
17957914 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17929834 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG61572010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24305-63-3
Molekylformel C21 H36 O3
Molekylvikt (g/mol) 336.51
Synonym 9,12-Octadecadienoic acid (9Z,12Z)-, oxiranylmethyl ester (9CI), 9,12-Octadecadienoic acid (Z,Z)-, oxiranylmethyl ester, Linoleic acid, 2,3-epoxypropyl ester (8CI), 1-Propanol, 2,3-epoxy-, linoleate (8CI), 2,3-Epoxypropyl linoleate, Glycidyl linoleate, Linoleic acid glycidyl ester, 9,12-Octadecadienoic acid (9Z,12Z)-, 2-oxiranylmethyl ester, Linoleic acid-glycidyl ester
IUPAC-namn oxiran-2-ylmethyl (9Z,12Z)-octadeca-9,12-dienoate
LEDER CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC1CO1
Specifikationer
Kemiskt namn eller material Glycidyl Linoleate
Anteckningar om renhetsgrad GC
Kvantitet 10 mg
InChI formel InChI=1S/C21H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h6-7,9-10,20H,2-5,8,11-19H2,1H3/b7-6-,10-9-
Formel vikt 336.2664
Procent renhet >95%