Läs mer
Glycidyl Stearate, TRC
Produktkod. 30009185
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009185 2.5 g 2.5g
17957694 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG6159852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7460-84-6
Molekylformel C21 H40 O3
Molekylvikt (g/mol) 340.54
Synonym Glycidyl Stearate, Octadecanoic acid, 2-oxiranylmethyl ester, Octadecanoic acid, oxiranylmethyl ester (9CI), Stearic acid, 2,3-epoxypropyl ester (6CI,7CI,8CI), 1-Propanol, 2,3-epoxy-, stearate (8CI), Glycidol stearate, Glycidyl octadecanoate, Glycidyl stearate, NSC 404228, Stearic acid glycidyl ester
IUPAC-namn oxiran-2-ylmethyl octadecanoate
LEDER CCCCCCCCCCCCCCCCCC(=O)OCC1CO1
Specifikationer
Kemiskt namn eller material Glycidyl Stearate
Anteckningar om renhetsgrad GC
Kvantitet 2.5 g
InChI formel InChI=1S/C21H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h20H,2-19H2,1H3
Formel vikt 340.2977
Procent renhet >95%