Läs mer
Glycine Benzyl Ester Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Glycine Benzyl Ester |
| CAS | 2462-31-9 |
| Molekylformel | C9 H11 N O2 . Cl H |
| Synonym | Glycine Phenylmethyl Ester Hydrochloride (1:1);Glycine, benzyl ester, hydrochloride (6CI,7CI,8CI), Glycine, phenylmethyl ester, hydrochloride (9CI), Aminoacetic acid benzyl ester hydrochloride, Benzyl 2-aminoacetate hydrochloride, Benzyl glycinate hydrochloride, Glycine benzyl ester monohydrochloride |
| InChI formel | InChI=1S/C9H11NO2.ClH/c10-6-9(11)12-7-8-4-2-1-3-5-8;/h1-5H,6-7,10H2;1H |
| LEDER | Cl.NCC(=O)OCc1ccccc1 |
| IUPAC-namn | benzyl 2-aminoacetate;hydrochloride |
| Molekylvikt (g/mol) | 201.65 |
| Kvantitet | 5 g |
| Formel vikt | 201.0557 |
| Visa mer |