Läs mer
Glycinexylidide Hydrochloride, TRC
Produktkod. 30008796
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008796 1 g
30003542 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008796 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG6252901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35891-83-9
Molekylformel C10 H14 N2 O . Cl H
Molekylvikt (g/mol) 214.69
Synonym Acetamide, 2-amino-N-(2,6-dimethylphenyl)-, hydrochloride (1:1), Acetamide, 2-amino-N-(2,6-dimethylphenyl)-, monohydrochloride (9CI), Glycine 2,6-xylidide hydrochloride
LEDER Cl.Cc1cccc(C)c1NC(=O)CN
Specifikationer
Kemiskt namn eller material Glycinexylidide Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C10H14N2O.ClH/c1-7-4-3-5-8(2)10(7)12-9(13)6-11;/h3-5H,6,11H2,1-2H3,(H,12,13);1H
Formel vikt 214.0873
Procent renhet >95%