Läs mer
Glycodeoxycholic Acid Sodium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Glycodeoxycholic Acid Sodium Salt |
| CAS | 16409-34-0 |
| Molekylformel | C26 H42 N O5 . Na |
| Kvantitet | 1 g |
| Synonym | Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, sodium salt (1:1), Glycine, N-(3α,12α-dihydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, monosodium salt (9CI), Glycodeoxycholic acid, sodium salt (6CI,7CI), Cholane, glycine deriv., Sodium deoxyglycolate, Sodium glycodeoxycholate, Sodium glycyldeoxycholate, Glycodeoxycholic acid sodium salt, N-[(3α,5β,12α)-3,12-Dihydroxy-24-oxocholan-24-yl]glycine monosodium salt |
| InChI formel | InChI=1S/C26H43NO5.Na/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3;/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;/m1./s1 |
| LEDER | [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC-namn | sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate |
| Molekylvikt (g/mol) | 471.61 |
| Formel vikt | 471.2961 |