Läs mer
Glycolaldehyde-1,2-13C2 (0.1 M in water), TRC
Produktkod. 30179971
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179971 50 mg
30179939 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG64110450MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 478529-69-0
Molekylformel 13C2 H4 O2
Molekylvikt (g/mol) 62.04
Synonym 2-Hydroxyacetaldehyde-1,2-13C2, 2-Hydroxyethanal-1,2-13C2, 2-Oxoethanol-1,2-13C2, Hydroxyacetaldehyde-1,2-13C2, Diose-1,2-13C2, Glicolaldehido-1,2-13C2, Glycolic Aldehyde-1,2-13C2, Glycollaldehyde-1,2-13C2, Hydroxyethanal-1,2-13C2, Methylolformaldehyde-1,2-13C2, Monomethylolformaldehyde-1,2-13C2, NSC 67935-1,2-13C2
IUPAC-namn 2-hydroxyacetaldehyde
LEDER O[13CH2][13CH]=O
Specifikationer
Kemiskt namn eller material Glycolaldehyde-1,2-13C2
Lösningsmedel eller matris Water
Kvantitet 50 mg
InChI formel InChI=1S/C2H4O2/c3-1-2-4/h1,4H,2H2/i1+1,2+1
Formel vikt 62.0278