Läs mer
Glycolic Acid-13C2, TRC
Produktkod. 30133699
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133699 10 mg
30135427 50 mg
17903885 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG64560210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 111389-68-5
Molekylformel 13C2H4O3
Molekylvikt (g/mol) 78.04
Synonym 2-Hydroxyacetic Acid-13C2, 2-Hydroxyethanoic Acid-13C2, GlyPure-13C2, GlyPure 70-13C2, GlyPure 99-13C2, Glycocide-13C2, Hydroxyacetic Acid-13C2, Hydroxyethanoic Acid-13C2, NSC 166-13C2, α-Hydroxyacetic Acid-13C2;
LEDER O[13CH2][13C](O)=O
Specifikationer
Kemiskt namn eller material Glycolic Acid-13C2
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)/i1+1,2+1
Formel vikt 78.0228
Procent renhet >95%