Learn More
Glycolic acid, Sigma-Aldrich™
Product Code. 32238507
Change view
Click to view available options
Quantity:
25 L
Unit Size:
25 L
Product Code. Quantity Unit Size
32238507 25 L 25 L
1 options
Product Code. 32238507 Supplier Sigma Aldrich Supplier No. 8146629026
 Restricted Item
Beräknad leverans: 26-11-2026
Add to basket
Add to basket
(70% solution in water) special quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Glycolic acid
pH 0.5 (20°C, 700 g/L in H2O)
Melting Point 10°C
Boiling Point 112°C (1013 hPa)
Density 1.26 g/cm³ at 20°C
Quantity 25 L
Grade Not Available