Läs mer
Glycolithocholic Acid Sodium Salt, TRC
Produktkod. 30004561
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004561 10 mg
30009062 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG64565310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24404-83-9
Molekylformel C26H42NNaO4
Molekylvikt (g/mol) 455.61
Synonym Sodium Glycolithocholate, N-(3alpha-Hydroxy-5beta-cholan-24-oyl)glycine Sodium Salt, N-[(3alpha,5beta)-3-Hydroxy-24-oxocholan-24-yl]glycine Sodium Salt;
IUPAC-namn sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
LEDER [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Kemiskt namn eller material Glycolithocholic Acid Sodium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C26H43NO4.Na/c1-16(4-9-23(29)27-15-24(30)31)20-7-8-21-19-6-5-17-14-18(28)10-12-25(17,2)22(19)11-13-26(20,21)3;/h16-22,28H,4-15H2,1-3H3,(H,27,29)(H,30,31);/q;+1/p-1/t16-,17-,18-,19+,20-,21+,22+,25+,26-;/m1./s1
Formel vikt 455.3012