missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycoluril, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300.0°C |
Boiling Point | 862.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (N) |
Quantity | 25 g |
Beilstein | 26,441 |
Merck Index | 15,88 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in warm mineral acids and in warm ammonia |
Formula Weight | 142.12 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- Glycoluril
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'