Läs mer
Glycopyrrolate Bromide, TRC
Produktkod. 30004544
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004544 10 mg
30004952 5 mg
30006633 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG65698010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 596-51-0
Molekylformel C19 H28 N O3 . Br
Molekylvikt (g/mol) 398.33
Synonym Pyrrolidinium, 3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethyl-, bromide (1:1), 3-Hydroxy-1,1-dimethylpyrrolidinium bromide α-cyclopentylmandelate (6CI,7CI), Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide (9CI), Pyrrolidinium, 3-hydroxy-1,1-dimethyl-, bromide, α-cyclopentylmandelate (8CI), 1,1-Dimethyl-3-hydroxypyrrolidinium bromide α-cyclopentylmandelate, AHR-504, Asecryl, Copyrrolate, Gastrodyn, Glycopyrrolate, Glycopyrrolate bromide, Glycopyrrone bromide, Glycopyrronium bromide, NSC 250836, NSC 251251, NSC 251252, NVA 237, Nodapton, Robanul, Robinul, Seebri, Tarodyl, Tarodyn, β-1-Methyl-3-pyrrolidyl-α-cyclopentylmandelate methobromide
IUPAC-namn (1,1-dimethylpyrrolidin-1-ium-3-yl) 2-cyclopentyl-2-hydroxy-2-phenylacetate;bromide
LEDER [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C2CCCC2)c3ccccc3
Specifikationer
Kemiskt namn eller material Glycopyrrolate Bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1
Formel vikt 397.1253
Procent renhet >95%