Läs mer
Glycoursodeoxycholic Acid Sodium Salt, TRC
Produktkod. 17997614
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17997614 250 mg
17979974 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17997614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG679545250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 92411-07-9
Molekylformel C26H42NNaO5
Molekylvikt (g/mol) 471.61
Synonym N-[(3.alpha.,5.beta.,7.beta.)-3,7-dihydroxy-24-oxocholan-24-yl]Glycine Sodium Salt, 3.alpha.,7.beta.-Dihydroxy-5.beta.-cholanoylglycine, Glycylursodeoxycholic acid Sodium Salt, N-(3.alpha.,7.beta.-Dihydroxy-5.beta.-cholan-24-oyl)glycine Sodium Salt, N-[(3.alpha.,5.beta.,7.beta.)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium Salt, Ursodeoxycholylglycine Sodium Salt, Glycoursodiol Sodium Salt
IUPAC-namn sodium;2-[[(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
LEDER [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Glycoursodeoxycholic Acid Sodium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21+,24+,25+,26-;/m1./s1
Formel vikt 471.2961