missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GLYCTK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582268
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: IRAAMERAGK QEMLLKPHSR VQVFEGAEDN LPDRDALRAA LAIQQLAEGL TADDLLLVLI SGGGSALLPA PIPPVTLEEK QTLTRLLAAR GATIQELNTI RKALSQLKGG GLAQAAYPAQ VVSLILSDVV G Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifikationer
| GLYCTK | |
| Polyclonal | |
| Unconjugated | |
| GLYCTK | |
| 6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 132158, 235582 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVS8, Q8QZY2 | |
| GLYCTK | |
| Recombinant protein corresponding to Human GLYCTK. Recombinant protein control fragment (Product #RP-89448). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering