Läs mer
Glycyl-DL-alanine, TRC
Produktkod. 30404349
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30404349 250 mg
1 options
Produktkod. 30404349 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG115350250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Glycyl-DL-alanine
CAS 926-77-2
Molekylformel C5H10N2O3
Kvantitet 250 mg
Synonym Glycylalanine, 2-(2-Aminoacetamido)propanoic Acid, 2-[(2-Azaniumylacetyl)amino]propanoate, Gly-DL-Ala, Glycyl-(RS)-alanine, Glycyl-DL-alanine, Glycyl-DL-alpha-alanine
InChI formel InChI=1S/C5H10N2O3/c1-3(5(9)10)7-4(8)2-6/h3H,2,6H2,1H3,(H,7,8)(H,9,10)
LEDER CC(NC(=O)CN)C(=O)O
IUPAC-namn 2-[(2-aminoacetyl)amino]propanoic acid
Molekylvikt (g/mol) 146.14
Formel vikt 146.0691