Läs mer
Glyoxal (40% w/w in H2O) (Technical Grade), TRC
Produktkod. 30012846
Change view
Kvantitet:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012846 25 mL
30138407 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012846 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG75450025ML
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 107-22-2
Kemiskt namn eller material Glyoxal
Formel vikt 58.0055
InChI formel InChI=1S/C2H2O2/c3-1-2-4/h1-2H
IUPAC-namn oxaldehyde
Molekylformel C2H2O2
Molekylvikt (g/mol) 58.04
LEDER O=CC=O
Synonym Ethanedial, Glyoxal (8CI), 1,2-Ethanedione, Aurarez 136, Biformal, Biformyl, Cartabond GH, Cartabond GHF, Daicel GY 60, Diformyl, Earth Works Linkup Plus, Ethanedione, Freechem 40DL, GX, GX (aldehyde), Glyfix CS 50, Glyoxal 40L, Glyoxal T 40, Glyoxal aldehyde, Glyoxazal, Glyoxazal GX, Glyoxylaldehyde, Gohsezal P, Oxal, Oxalaldehyde, Permafresh 114, Protorez BLF-C, XH 536
Specifikationer
Kvantitet 25 mL