Learn More
Glyoxal dimethyl acetal, 60% aq. soln.
Product Code. 11335015
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335015 2000 g 2kg
11315015 100 g 100g
11325015 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11335015 Supplier Thermo Scientific Alfa Aesar Supplier No. L18490.0P
 Restricted Item
I lager
Add to basket
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51673-84-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00134410
InChI Key OGFKTAMJLKHRAZ-UHFFFAOYSA-N
Synonym acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
PubChem CID 162650
IUPAC Name 2,2-dimethoxyacetaldehyde
SMILES COC(C=O)OC
Specifications
Density 1.15
Boiling Point 100°C to 102°C
Flash Point 102°C (216°F)
Refractive Index 1.414
Quantity 2000 g
Beilstein 1850744
Solubility Information Fully miscible in water.
Formula Weight 104.11
Concentration or Composition (by Analyte or Components) 60% aq. soln.
Chemical Name or Material Glyoxal dimethyl acetal