Läs mer
Glyoxylic Acid-13C2 Monohydrate, TRC
Produktkod. 30134798
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134798 10 mg
30006350 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG75500210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-99-8
Molekylformel 13C2H4O4
Molekylvikt (g/mol) 94.04
Synonym 2-Oxoacetic Acid-13C2 Monohydrate, Glyoxalic Acid-13C2 Monohydrate, Formylformic Acid-13C2 Monohydrate, NSC 27785-13C2, Oxalaldehydic Acid-13C2 Monohydrate, Oxoacetic Acid-13C2 Monohydrate, Oxoethanoic Acid-13C2 Monohydrate, α-Ketoacetic Acid-13C2 Monohydrate;
IUPAC-namn 2-oxoacetic acid;hydrate
LEDER O=[13CH][13C](O)=O.O
Specifikationer
Kemiskt namn eller material Glyoxylic Acid-13C2 Monohydrate
Kvantitet 10 mg
InChI formel InChI=1S/C2H2O3.H2O/c3-1-2(4)5;/h1H,(H,4,5);1H2/i1+1,2+1;
Formel vikt 94.0177