missing translation for 'onlineSavingsMsg'
Läs mer

Glyphosate Isopropylamine Salt, TRC

Produktkod. 30004193
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004193 50 mg
17993975 10 mg
17951325 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30004193 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG76500450MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 38641-94-0
Molekylformel C3 H9 N . C3 H8 N O5 P
Molekylvikt (g/mol) 228.18
Synonym Glycine, N-(phosphonomethyl)-, compd. with 2-propanamine (1:1), 2-Propanamine, compd. with N-(phosphonomethyl)glycine (1:1), Accord concentrate (herbicide), Amega, AquaMaster, Atut, Azural AT, Bayer Glyphosate Concentrate, Buggy, Cargly, Ceresate, Clinic Ace, Clinic E.V., Corphosate, Cosmic, Cosmic (herbicide), Credit, Cuspide 480SL, Durango, Eraser (herbicide), Excel Mera 71, Fakel, Fosulen, Gallup 360SL, Glifocyd, Glifogan, Glifosato Atanor, Glyfos AU, Glyfos BIO, Glyphogan, Glyphogan Classic, Glyphomax Plus, Glyphomax XRT, Glyphosate AKB 480, Glyphosate AKB 80, Glyphosate Atanor 48, Glyphosate IPA salt, Glyphosate isopropylamine, Glyphosate isopropylamine salt, Glyphosate isopropylammonium salt, Glyphosate mono(isopropylamine) salt, Glypro, Glypro Bio, Glystar Plus, GrowChoice, Helosate 70, Helosate Plus, Ken-Up, Klinik Duo, MON 0139, MON 39, N-(Phosphonomethyl)glycine isopropylamine salt, N-(Phosphonomethyl)glycine isopropylammonium salt, N-(Phosphonomethyl)glycine monoisopropylamine salt, Nitosorg, Ortho Basic Solutions weed & grass killer Ready-to-Use, Otaman, Panzer, Pratiko, Razor, Razor Pro, Razor Pro Herbicide 4467, Rodeo, Ron-do, Roundup, Roundup 3 plus, Roundup Biactive, Roundup Bio, Roundup Biovert, Roundup Custom, Roundup Flash, Roundup High Load, Roundup LB Plus, Roundup Original, Roundup Superconcentrate, Roundup Transorb, Roundup Turbo, Roundup Ultra, Roundup UltraMax, Sangle, Spectra, Sun-Fulon, Topglypho 360, Transorb, Trop, Utal, Utal (herbicide), Vision, Vision (herbicide), Vulkan, Zapp
IUPAC-namn 2-(phosphonomethylamino)acetic acid;propan-2-amine
LEDER CC(C)N.OC(=O)CNCP(=O)(O)O

Specifikationer

Kemiskt namn eller material Glyphosate, isopropylamine salt
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C3H8NO5P.C3H9N/c5-3(6)1-4-2-10(7,8)9;1-3(2)4/h4H,1-2H2,(H,5,6)(H2,7,8,9);3H,4H2,1-2H3
Formel vikt 228.0875
Procent renhet >95%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.