missing translation for 'onlineSavingsMsg'
Learn More

Glypican 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16700-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886414

  • 3665.00 SEK / 150µL
Estimated Shipment: 22-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glypican 1 is a cell surface heparan sulfate proteoglycan that is composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
Specifications

Specifications

Glypican 1
Polyclonal
Unconjugated
GPC1
glypican, Glypican 1, GPC1
Rabbit
Antigen Affinity Chromatography
RUO
14733, 2817, 58920
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35052, P35053, Q9QZF2
Gpc1
Glypican 1 Fusion Protein Ag10095
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers