Learn More
Glypican 4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13048-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16871064
  • 4721.67 SEK / 150µL
Estimated Shipment: 30-04-2024
to see stock.
Description
Description
The Glypican 4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Glypican 4. This antibody recognizes human, mouse, rat antigen. The Glypican 4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Glypican 4
Polyclonal
Unconjugated
150 μL
2239
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
240 μg/mL
Rabbit
Primary
Human, Rat, Mouse