Learn More
Glypican 4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13048-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The Glypican 4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Glypican 4. This antibody recognizes human, mouse, rat antigen. The Glypican 4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Glypican 4 | |
Polyclonal | |
Unconjugated | |
150 μL | |
2239 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
240 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |