missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
GM-CSF Rat, Clone: BVD2-21C11, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-32967-0.1MG
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Neutralization.
Specifications
| GM-CSF | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| Rat | |
| 22 kDa | |
| 0.1 mg | |
| Cytokine Research, Immunology | |
| 1437 | |
| Human, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| BVD2-21C11 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate | |
| P04141 | |
| CSF2 | |
| Recombinant human GM-CSF protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors. | |
| Store at 4C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction