Learn More
Multiroir Controlec™ GM Sectioned Drawer Inserts
Fits securely in the tray of a GM Drawer Cabinet. Multiroir Controlec™ GM Sectioned Drawer Inserts are thermoformed honeycomb grid pieces that customize the plastic drawer with the size and number of compartments needed for sectioned storage.
Brand: Multiroir Controlec™ 32155
Additional Details : Weight : 0.15000kg
Description
GM Drawer Cabinets are a block of six colored tray-drawers to suit a variety of sorted storage needs.
- Solid polypropylene construction
- Color coded trays / drawers
- Transportable and stackable
- Drawers can be equipped with a variety of honeycomb grids
- Drawer/Insert Size: 340 x 250 x 40mm
Specifications
Plastic | |
Sectioned storage | |
40 mm | |
GM Drawer Cabinets | |
77 mm (compartment) |
Sectioned Drawer Insert | |
16 Compartments | |
White | |
55 mm (compartment) | |
4 x 4 |