Learn More
GMEB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552678
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSVPLGKVVS TLPSTVLGKG SLQAPPASSP ASPLLGGYTV LASSGSTYPS TVEIHPDASS LTVLSTAAVQ DGSTVFKVVS PLQLLTLPGL GPTLQNVAQA SPGSSTIVTV PAGAAPGPEE HTATIE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
GMEB2 Polyclonal antibody specifically detects GMEB2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GMEB2 | |
Polyclonal | |
Unconjugated | |
GMEB2 | |
AI839884; DNA-binding protein p79PIF; glucocorticoid modulatory element binding protein 2; glucocorticoid modulatory element-binding protein 2; GMEB2; GMEB-2; KIAA1269; P79PIF; parvovirus initiation factor p79; parvovirus initiation factor, p79; PIF p79; PIF79 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKD1 | |
GMEB2 | |
Recombinant protein corresponding to Human GMEB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only