missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMIP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25917-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing zinc finger domains. A short peptide derived from this protein is a minor histocompatibility antigen which can lead to graft rejection of male donor cells in a female recipient. Alternative splicing results in multiple transcript variants.Specifications
GMIP | |
Polyclonal | |
Unconjugated | |
GMIP | |
GEM interacting protein, GMIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51291 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P107 | |
GMIP | |
GMIP Fusion Protein Ag23239 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |