missing translation for 'onlineSavingsMsg'
Läs mer

AcroBiosystems GMP Recombinant Human Fibronectin fragment

Produktkod. 31452588 Handla allt ACROBiosystems Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
31452588 10 mg
31452901 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 31452588 Leverantör AcroBiosystems Leverantörsnummer GMPFINH1810mg
 Begränsad artikel
Beräknad leverans: 12-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Fibronectin (Fn) is a glycoprotein whose size ranges from 230 to 270 kDa and usually exists as a dimer, covalently linked by a pair of disulfide bonds at the C-termini. Each monomer consists of three repeating units: 12 Type I, 2 Type II, and 15-17 Type III domains which combined account for 90% of the FN sequence. The extracellular matrix (ECM) plays a key role as both structural scaffold and regulator of cell signal transduction in tissues. Fibronectin is one of the major ECM proteins in the trabecular meshwork (TM). It is found in the sheath material surrounding the elastin tendons that enter the TM from the ciliary muscle within the ciliary body. In times of ECM assembly and turnover, cells upregulate assembly of the ECM protein, FN. FN is assembled by cells into viscoelastic fibrils that can bind upward of 40 distinct growth factors and cytokines. These fibrils play a key role in assembling a provisional ECM during embryonic development and wound healing. Fibril assembly is also often upregulated during disease states, including cancer and fibrotic diseases.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer Uniprot
Karakterisering Fibronectin Structure This protein carries no tag. The protein has a calculated MW of 62.6 kDa. The protein migrates as 60 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE).
Formatera Liquid
Formulering Supplied as 0.2 μm filtered solution in 12.5 mM Citric acid, pH 6.2 with protectants. Contact us for customized product form or formulation.
Kvalitetskontrolltestning <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Kvantitet 10 mg
Källa GMP Recombinant Human Fibronectin fragment (GMP-FINH18) is expressed from E. coli cells. It contains AA Pro 1361 - Ser 1637 & Ala 1812 - Thr 2107 (Accession # ). Predicted N-terminus: Met Request for sequence
Biologisk aktivitet Biotinylated Human ITGA4&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT1-H82W1) immobilized on SA Chip can bind GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) with a typical affinity constant of 0.799 μM as determined in a SPR assay (Biacore 8K) (QC tested).∣Biotinylated Human ITGA5&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT1-H82Wa) immobilized on SA Chip can bind GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) with a typical affinity constant of 35.7 nM as determined in a SPR assay (Biacore 8K) (QC tested).∣GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) immobilized on CM5 Chip can bind Heparin Sulfate with a typical affinity constant of 63.7 nM as determined in a SPR assay (Biacore 8K) (QC tested).∣GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) could promote mesenchymal stem cell attachment and proliferation in several passages, comparable with Competitor T. The bioactivity of GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is similar to Recombinant Fibronectin fragment, premium grade (Cat. No. FIN-H5116).∣GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) could maintain the stemness of MSC in several passages, with high expression of MSC markers CD73, CD90 and CD105 and negative expression of HLA-DR, CD34 and CD45. The stemness ability of GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is comparable with Competitor T and similar to Recombinant Fibronectin fragment, premium grade (Cat. No. FIN-H5116).∣CD34+ CD45+ hematopoietic cells were seeded on GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) combined with GMP Human DLL4 Protein, Fc Tag (Flagship) (Cat. No. GMP-DL4H27) and GMP Human VCAM-1 Protein, Fc Tag (Cat. No. GMP-VC1H25) coated plates and differentiated for 21 days, then flow cytometry was used to detect the expression of T-cell progenitor markers, CD5 and CD7. GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) combined with GMP Human DLL4 Protein, Fc Tag (Flagship) (Cat. No. GMP-DL4H27) and GMP Human VCAM-1 Protein, Fc Tag (Cat. No. GMP-VC1H25) can induce the formation of CD7+ and CD5+ CD7+ T-cell progenitors. The bioactivity of GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is similar to Recombinant Fibronectin fragment, premium grade (Cat. No. FIN-H5116), as well as the competitor T. Negative control: w/o Human Fibronectin.∣The SPR based assay shows that GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is stable at 37°C for 23 days.∣The SPR based assay shows that GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is stable at 4°C for 180 days.∣The SPR based assay shows that GMP Recombinant Human Fibronectin fragment (Cat. No. GMP-FINH18) is stable after freezing and thawing 3 times.
Art Human
Sterilitet Sterile
Innehåll och lagring -20°C for 3 years under sterile conditions.
Endotoxinnivå Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Frakt skick This product is supplied and shipped with dry ice, please inquire the shipping cost.
Renhet eller kvalitetsklass 0.95
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.