Läs mer
Tocris Bioscience™ GN 44028
Potent HIF-1α inhibitor
Brand: Tocris Bioscience™ 5655/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GN 44028 is a potent HIF-1α inhibitor (IC50 = 14 nM). Inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. Cytotoxic to a range of cancer cell types in vitro.Specifikationer
| N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine | |
| 1421448-26-1 | |
| 10 mg |