missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ GNAQ (Human) Recombinant Protein

Human GNAQ full-length ORF ( AAH57777, 1 a.a. - 359 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002776-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115561

  • 4055.00 SEK / 10µg
Estimated Shipment: 05-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MTLESIMACCLSEEAKEARRINDEIERQLRRDKRDARRELKLLLLGTGESGKSTFIKQMRIIHGSGYSDEDKRGFTKLVYQNIFTAMQAMIRAMDTLKIPYKYEHNKAHAQLVREVDVEKVSAFENPYVDAIKSLWNDPGIQECYDRRREYQLSDSTKYYLNDLDRVADPAYLPTQQDVLRVRVPTTGIIEYPFDLQSVIFRMVDVGGQRSERRKWIHCFENVTSIMFLVALSEYDQVLVESDNENRMEESKALFRTIITYPWFQNSSVILFLNKKDLLEEKIMYSHLVDYFPEYDGPQRDAQAAREFILKMFVDLNPDSDKIIYSHFTCATDTENIRFVFAAVKDTILQLNLKEYNLV
Specifications

Specifications

AAH57777
guanine nucleotide binding protein (G protein), q polypeptide
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GNAQ
GST
2776
Wheat germ expression system
10 μg
G-ALPHA-q, GAQ
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers