Läs mer
Tocris Bioscience™ GNE 2861
Beskrivning
GNE 2861 is an inhibitor of group II p21 activated kinases (PAK; IC50 values are 7.5, 126 and 36 nM for PAK4, PAK5 and PAK6, respectively). Displays selectivity for group II PAK over a panel of 222 kinases. Inhibits tumor cell proliferation, migration and invasion in two breast cancer cell lines. Also sensitizes tamoxifen-resistant breast cancer cell lines to tamoxifen (Cat. No. 6342) via PAK4-dependent regulation of estrogen receptor α (ERα) target gene expression.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-[2-[1-(2-Amino-4-pyrimidinyl)-2-[(2-methoxyethyl)amino]-1H-benzimidazol-6-yl]ethynyl]cyclohexanol |
| CAS | 1394121-05-1 |
| Kvantitet | 5 mg |
| Molekylformel | C22H26N6O2 |
| Formel vikt | 406.48 |
| Renhet | 98% |