Läs mer
GNE-3511, MedChemExpress
Produktkod. 16491617
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
Förpackningsstorlek:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16491617 50 mg -
16456036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16491617 Leverantör Medchem Express Leverantörsnummer HY12947
MedChemExpress GNE-3511 is an orally active bioavailable and brain-penetrant dual leucine zipper kinase (DLK) inhibitor with a Ki of 0.5 nM. GNE-3511 can cross the blood-brain-barrier and can be used for the research of neurodegenerative diseases.
GNE-3511 is an orally active bioavailable and brain-penetrant dual leucine zipper kinase (DLK) inhibitor with a Ki of 0.5 nM. GNE-3511 can cross the blood-brain-barrier and can be used for the research of neurodegenerative diseases.
Specifikationer
Kemiskt namn eller material GNE-3511
CAS 1496581-76-0
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C23H26F2N6O
Kvantitet 50 mg
Löslighetsinformation DMSO : 31.25 mg/mL (70.94 mM; Need ultrasonic)
LEDER N#CC1=CC=NC(NC2=NC(N3CC(F)(F)CC3)=CC(C4CCN(C5COC5)CC4)=C2)=C1
Molekylvikt (g/mol) 440.49
Formel vikt 440.49
Procent renhet 98.87%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease