Learn More
GNE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25079-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of maligt cells.Specifications
GNE | |
Polyclonal | |
Unconjugated | |
GNE | |
DMRV, GLCNE, GNE, IBM2, ManAc kinase, NM, Uae1, UDP GlcNAc 2 epimerase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10020 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y223 | |
GNE | |
GNE Fusion Protein Ag18689 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |