Learn More
GNF-5837, MedChemExpress
Product Code. 16491867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491867 50 mg 50mg
16486196 100 mg 100mg
16475708 10 mg 10mg
16408407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16491867 Supplier Medchem Express Supplier No. HY13491
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress GNF-5837 is a potent, selective, and orally bioavailable pan-tropomyosin receptor kinase (TRK) inhibitor which display antiproliferative effects in cellular Ba/F3 assays ( IC50 values of 7 nM, 9 nM and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively) .
GNF-5837 is a potent, selective, and orally bioavailable pan-tropomyosin receptor kinase (TRK) inhibitor which display antiproliferative effects in cellular Ba/F3 assays ( IC50 values of 7 nM, 9 nM and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively) .
Specifications
Chemical Name or Material GNF-5837
CAS 1033769-28-6
Color Orange
Purity Grade Notes Research
Molecular Formula C28H21F4N5O2
Quantity 50 mg
Solubility Information DMSO : ≥ 32 mg/mL (59.76 mM)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1F)NC2=CC=C(C)C(NC3=CC(NC/4=O)=C(C=C3)C4=C\C5=CC=CN5)=C2
Molecular Weight (g/mol) 535.49
Formula Weight 535.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease