missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNG4 Monoclonal Antibody (1D9-G11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524407
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: MKEGMSNNST ASISQARKAV EQLKMEACMD RVKVSQAAAD LLAYCEAHVR EDPLIIPVPA SENPFREKKF FCTIL
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.Specifications
GNG4 | |
Monoclonal | |
Unconjugated | |
GNG4 | |
GNG4; GNGT4 | |
Mouse | |
Protein A | |
RUO | |
2786 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1D9-G11 | |
PBS with no preservative; pH 7.4 | |
P50150 | |
GNG4 | |
GNG4 (AAH22485, 1 a.a. ∽ 75 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |