missing translation for 'onlineSavingsMsg'
Learn More

GNL2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10190-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876403

  • 4721.67 SEK / 150µL
Estimated Shipment: 25-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation.
Specifications

Specifications

GNL2
Polyclonal
Unconjugated
GNL2
Autoantigen NGP 1, GNL2, HUMAUANTIG, Ngp 1, NGP1
Rabbit
Antigen Affinity Chromatography
RUO
29889
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13823
GNL2
GNL2 Fusion Protein Ag0243
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers