missing translation for 'onlineSavingsMsg'
Learn More

GNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18790-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878624

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
Specifications

Specifications

GNMT
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13255, Q14749, Q9QXF8
glycine N methyltransferase, GNMT
Rabbit
IgG
20 μL
-20°C
Primary
14711, 25134, 27232
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
GNMT
Liquid
GNMT
GNMT Fusion Protein Ag4598
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers