missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GNMT |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878624
|
Proteintech
18790-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868624
|
Proteintech
18790-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.Specifications
GNMT | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P13255, Q14749, Q9QXF8 | |
14711, 25134, 27232 | |
GNMT Fusion Protein Ag4598 | |
Primary | |
-20°C | |
GNMT |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
glycine N methyltransferase, GNMT | |
GNMT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |