Läs mer
ABCC13, Goat, Polyclonal Antibody, Abnova™
Beskrivning
This gene is a member of the superfamily of genes encoding ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This family member is part of the MRP subfamily, which is involved in multi-drug resistance, but the human locus is now thought to be a pseudogene incapable of encoding a functional ABC protein. Alternative splicing results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | ABCC13 |
| Användningsområden | ELISA |
| Klassificering | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Goat polyclonal antibody raised against synthetic peptide of ABCC13. |
| Utspädning | ELISA (1:128000) The optimal working dilution should be determined by the end user. |
| Formulering | In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) |
| Gen | ABCC13 |
| Genaccessionsnr. | AAL47201.1;AAQ22368.1;EAX10059.1 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.