Läs mer
ANXA2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ANXA2.
Brand: Abnova PAB6109.100ug
Beskrivning
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: STVHEILCKLSLEGDSpecifikationer
| ANXA2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANXA2 | |
| ANX2/ANX2L4/CAL1H/LIP2/LPC2/LPC2D/P36/PAP-IV | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 302 | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ANXA2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001002858.1;NP_004030.1;NP_001002857.1 | |
| ANXA2 | |
| A synthetic peptide corresponding to human ANXA2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms. NP_004030.1 and NP_001002857.1 are varients of isoform 2. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only