missing translation for 'onlineSavingsMsg'
Läs mer

ARMET, Goat, Polyclonal Antibody, Abnova™

Produktkod. 16070478
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16070478 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16070478 Leverantör Abnova Leverantörsnummer PAB7400.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Goat polyclonal antibody raised against synthetic peptide of ARMET.

The protein encoded by this gene is localized in the endoplasmic reticulum (ER) and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of a specific mutation changing the previously numbered codon 50 from ATG to AGG, or deletion of that codon, has been reported in a variety of solid tumors. With the protein size correction, this codon is now identified as the initiation codon. [provided by RefSeq

Sequence: KFCREARGKENR

Specifikationer

Antigen ARMET
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Beskrivning Goat polyclonal antibody raised against synthetic peptide of ARMET.
Utspädning ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gen ARMET
Genaccessionsnr. NP_006001.2
Gene Alias ARP/MANF/MGC142148/MGC142150
Gensymboler ARMET
Värd art Goat
Immunogen A synthetic peptide corresponding to human ARMET.
Reningsmetod Antigen affinity purification
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7873
Målarter Human
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.