Learn More
BIRC7, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BIRC7.
Brand: Abnova PAB6033.100ug
Description
The protein encoded by this gene is a member of the family of inhibitor of apoptosis proteins (IAP) and contains a single copy of a baculovirus IAP repeat (BIR) as well as a RING-type zinc finger domain. The BIR domain is essential for inhibitory activity and interacts with caspases, while the RING finger domain sometimes enhances antiapoptotic activity but does not inhibit apoptosis alone. Two transcript variants encoding different isoforms have been found for this gene. The two isoforms have different antiapoptotic properties, with isoform alpha protecting cells from apoptosis induced by staurosporine and isoform b protecting cells from apoptosis induced by etoposide. [provided by RefSeq
Sequence: C-RAPVRSRVRTSpecifications
| BIRC7 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (0.1-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| BIRC7 | |
| KIAP/LIVIN/ML-IAP/MLIAP/RNF50 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 79444 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of BIRC7. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_647478.1;NP_071444.1 | |
| BIRC7 | |
| A synthetic peptide corresponding to human BIRC7. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (as represented by NP_647478.1 and NP_071444.1) | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only