missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Goat anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, DyLight 594, R&D Systems™
Handla allt Bio Techne ProdukterDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Användningsområden | ELISA, Immunohistochemistry, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | DyLight 594 |
| Formulering | 50mM Sodium Borate |
| Värd art | Goat |
| Immunogen | Golden Syrian Hamster IgG whole molecule |
| Reningsmetod | Multi-step |
| Kvantitet | 0.5 mL |
| Regulatorisk status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?