Läs mer
HOXD12, Goat, Polyclonal Antibody, Abnova™
Beskrivning
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | HOXD12 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Goat polyclonal antibody raised against synthetic peptide of HOXD12. |
| Utspädning | ELISA (1:1000) Western Blot (2-3ug/ml) The optimal working dilution should be determined by the end user. |
| Formulering | In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) |
| Gen | HOXD12 |
| Gene Alias | HOX4H |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.