Läs mer
Goat anti-Human IgA Heavy Chain Secondary Antibody, Janelia Fluor 549, R&D Systems™
Handla allt Bio Techne ProdukterBeskrivning
IgA Heavy Chain Polyclonal specifically detects IgA Heavy Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.
Specifikationer
Specifikationer
| Antigen | IgA Heavy Chain |
| Användningsområden | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Janelia Fluor 549 |
| Formulering | 50mM Sodium Borate |
| Värd art | Goat |
| Immunogen | Highly purified human IgA alpha-chain |
| Reningsmetod | Affinity purified |
| Kvantitet | 0.1 mL |
| Regulatorisk status | RUO |
| Visa mer |