missing translation for 'onlineSavingsMsg'
Läs mer

Goat anti-Human IgG Fc Secondary Antibody, PE, Novus Biologicals™

Product Code. 30232397 Shop All Bio Techne Products
Change view
Click to view available options
Kvantitet:
0.5 mL
Unit Size:
0.5ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
30232397 0.5 mL 0.5ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30232397 Supplier Novus Biologicals Supplier No. NBP151846PE
 Begränsad artikel
Beräknad leverans: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Användningsområden ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Konjugera PE
Formulering PBS
Värd art Goat
Immunogen Human IgG Fc antiserum was solid phase adsorbed to ensure class specificity.
Reningsmetod Immunogen affinity purified
Kvantitet 0.5 mL
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Human
Innehåll och lagring Store at 4°C in the dark.
Form Purified
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.