Learn More
Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, Rhodamine, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31683
Additional Details : Weight : 0.80000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31683 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31683 reacts with the heavy chains of human IgG and with light chains common to most human immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, horse, and mouse serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Rhodamine Amax= 550 nm; Emax= 570 nm. Fluorophore/Protein Absorbance Ratio: A550/A280 = 0.44 (lot-dependent). Reconstitute with 1.1 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA
Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Specifications
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Rhodamine | |
Goat | |
1.5 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
1.5 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Human | |
Lyophilized |