Läs mer
Invitrogen™ Goat anti-Human IgG (H+L) Superclonal™ Secondary Antibody
Beskrivning
This Goat-anti-Human IgG (H+L), Superclonal™ Recombinant Secondary Antibody specifically detects human IgG Heavy and the Light chain with no cross-reactivity to other species immunoglobulins except moderate cross-reactivity towards Monkey IgG. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. Superclonal™ Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifikationer
Specifikationer
| Antigen | Human IgG (H+L) |
| Användningsområden | ELISA, Western Blot, Immunocytochemistry |
| Klassificering | Recombinant Superclonal |
| Koncentration | 1.0 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.09% sodium azide; pH 7.4 |
| Värd art | Goat |
| Immunogen | Purified Human IgG. |
| Reningsmetod | Protein G |
| Kvantitet | 250 μL |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.