missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgM Secondary Antibody, FITC, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen 31575

Additional Details : CAS Number : 17740-40-8 Weight : 0.00010kg

Product Code. 10341124

  • 1834.00 SEK / 2mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31575 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31575 reacts with the Fc5μ portion of human IgM heavy chain, but does not react against normal human IgG, IgA or against non-immunoglobulin serum proteins. However, this antibody may cross-react with IgM from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 9.0 μg/mg; > 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA

Human IgM Polyclonal antibody specifically detects Human IgM in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

Human IgM
Polyclonal
FITC
Goat
2 mg
Secondary
4° C, store in dark
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
1.5 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Affinity chromatography
RUO
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers