missing translation for 'onlineSavingsMsg'
Läs mer

Goat anti-Human Kappa Light Chain Secondary Antibody, HRP, R&D Systems™

Produktkod. 30228005 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30228005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30228005 Leverantör Novus Biologicals Leverantörsnummer NB7480H

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifica

Antigen Kappa Light Chain
Användningsområden Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Klassificering Polyclonal
Konjugera HRP
Formulering PBS
Gene Alias HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Värd art Goat
Immunogen Human Kappa Free Light Chain
Reningsmetod Immunogen affinity purified
Kvantitet 1 mL
Regulatorisk status RUO
Forskningsdisciplin Adaptive Immunity, Immunology
Primär eller sekundär Secondary
Gen-ID (Entrez) 3514
Målarter Human
Innehåll och lagring Store at 4°C in the dark.
Produkttyp Antibody
Form Purified
Isotyp IgG
Produktlinje Novus
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.