missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 594, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen R37121
Additional Details : Weight : 0.02000kg
Description
No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells.
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PBS with 0.1% BSA and <0.1% sodium azide | |
Gamma Immunoglobin | |
2 x 2.5 mL | |
Secondary | |
4° C, store in dark | |
IgG |
Flow Cytometry, Immunocytochemistry | |
Alexa Fluor 594 | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |