missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG Fc Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51898-0.5mg
2432.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
IgG Fc | |
Polyclonal | |
DyLight 650 | |
PBS containing 0.2% BSA | |
Mouse IgG | |
0.5 mg | |
Adaptive Immunity, Immunology | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
0.5 mg/mL | |
Flow Cytometry 1:50 - 1:200, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction